accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3HNOHNNOH3CHNOHNNOO

CAS RN 204067-01-6

IUPAC Name: 3-(1H-indol-3-yl)-N-{[1-(5-methoxypyridin-2-yl)cyclohexyl]methyl}-2-methyl-2-{[(4-nitrophenyl)carbamoyl]amino}propanamide
Synonyms: 3-(1H-indol-3-yl)-N-{[1-(5-methoxypyridin-2-yl)cyclohexyl]methyl}-2-methyl-2-{[(4-nitrophenyl)carbamoyl]amino}propanamide
Molecular Weight (g/mol): 584.68
Molecular Formula: C32H36N6O5
InChi Key: NNFUWNLENRUDHR-UHFFFAOYNA-N
SMILES: COC1=CN=C(C=C1)C1(CNC(=O)C(C)(CC2=CNC3=CC=CC=C23)NC(=O)NC2=CC=C(C=C2)[N+]([O-])=O)CCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)