CAS RN 2042192-00-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ Janelia Fluor 646, Tetrazine
Red dye (Reactive group: tetrazine). Applications: super-resolution microscopy e.g. dSTORM & STED, confocal and live cell imaging