accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2NNHOBrHOBrHNONNNHOONNN

CAS RN 204697-65-4

IUPAC Name: N-{6-amino-1-oxo-1-[4-(pyridin-4-yl)piperazin-1-yl]hexan-2-yl}-3-(3,5-dibromo-4-hydroxyphenyl)-2-{[4-(2-oxo-1,2,3,4-tetrahydroquinazolin-3-yl)piperidine-1-carbonyl]amino}propanamide
Synonyms: olcegepant
Molecular Weight (g/mol): 869.66
Molecular Formula: C38H47Br2N9O5
InChi Key: ITIXDWVDFFXNEG-UHFFFAOYNA-N
SMILES: NCCCCC(NC(=O)C(CC1=CC(Br)=C(O)C(Br)=C1)NC(=O)N1CCC(CC1)N1CC2=CC=CC=C2NC1=O)C(=O)N1CCN(CC1)C1=CC=NC=C1
Product Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)