Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ BIBN 4096
GREENER_CHOICESDP

Catalog No. 456110 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-611-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 45-611-0 Supplier Tocris Bioscience™ Supplier No. 4561/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective CGRP antagonist

Potent and selective CGRP receptor antagonist (IC50 values are 0.03 and 6.4 nM for human and rat CGRP receptors respectively). Displays high affinity for human CGRP receptors (Ki = 14.4 pM); exhibits no significant affinity for 75 other receptors.

Specifications

CAS 204697-65-4
Quantity 10 mg
Product Type BIBN 4096
Purity >96%
Molecular Weight (g/mol) 869.66
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.