accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONNNNHNClHOHOHO

CAS RN 205171-12-6

IUPAC Name: 2-[2-chloro-6-(cyclopentylamino)-9H-purin-9-yl]-5-(hydroxymethyl)-3-methyloxolane-3,4-diol
Synonyms: 2-[2-chloro-6-(cyclopentylamino)purin-9-yl]-5-(hydroxymethyl)-3-methyloxolane-3,4-diol
Molecular Weight (g/mol): 383.83
Molecular Formula: C16H22ClN5O4
InChi Key: MMPAUXMIDJWGFO-UHFFFAOYNA-N
SMILES: CC1(O)C(O)C(CO)OC1N1C=NC2=C(NC3CCCC3)N=C(Cl)N=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)