accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONNH2NNHN

CAS RN 20535-83-5

IUPAC Name: 6-methoxy-7H-purin-2-amine
Synonyms: 6-O-methylguanine
Molecular Weight (g/mol): 165.16
Molecular Formula: C6H7N5O
InChi Key: BXJHWYVXLGLDMZ-UHFFFAOYSA-N
SMILES: COC1=C2NC=NC2=NC(N)=N1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Grade 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results