Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-6-methoxypurine 97.0+%, TCI America™
SDP

Catalog No. A1151100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A1151100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A1151100MG Supplier TCI America Supplier No. A1151100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 20535-83-5
Molecular Formula C6H7N5O
Molecular Weight (g/mol) 165.156
MDL Number MFCD00133009
InChI Key BXJHWYVXLGLDMZ-UHFFFAOYSA-N
Synonym O-Methylguanine
PubChem CID 65275
ChEBI CHEBI:20689
IUPAC Name 6-methoxy-7H-purin-2-amine
SMILES COC1=NC(=NC2=C1NC=N2)N

Specifications

Color White
Quantity 100 mg
Formula Weight 165.16
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-6-methoxypurine
RTECSNumber UO7473000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.