CAS RN 2059395-52-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ Lomitapide mesylate
Potent microsomal triglyceride transfer protein (MTP) inhibitor