Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Lomitapide mesylate
GREENER_CHOICESDP

Catalog No. 749250 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
74-925-0 50 mg
74-921-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 74-925-0 Supplier Tocris Bioscience™ Supplier No. 7492/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent microsomal triglyceride transfer protein (MTP) inhibitor

Lomitapide mesylate is a potent inhibitor of microsomal triglyceride transfer protein (IC50 = 8 nM). Lomitapide inhibits or normalizes lipoprotein production in rodents and in a rabbit model of hypercholesterolemia. Lomitapide is antimalarial; it inhibits hemozoin formation and growth of CQ-sensitive (PfNF54) and multidrug-resistant (PfK1) strains of P. falciparum (IC50 values are 0.56 μM and 0.69 μM respectively). Orally bioavailable.

Specifications

Chemical Name or Material N-(2,2,2-Trifluoroethyl)-9-[4-[4-[[[4 '-(trifluoromethyl)[1,1 '-biphenyl]-2-yl]carbonyl]amino]-1-piperidinyl]butyl]-9 H-fluorene-9-carboxamide methanesulfonate
CAS 2059395-52-5
Quantity 50 mg
Target Lipid Metabolism Inhibitors
Molecular Formula C39H37F6N3O2.CH3SO3H
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.