CAS RN 2061980-01-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Tocris Bioscience™ XMU MP 1
Potent and selective ATP-competitive mammalian sterile 20-like kinase (MST) 1/2 inhibitor
2
Tocris Bioscience™ XMU MP 1
Potent and selective MST1/2 inhibitor