CAS RN 206996-13-6
IUPAC Name:
1-[2-(3,4-dichlorophenyl)ethyl]-4-methylpiperazine dihydrochloride
Synonyms:
1-[2-(3,4-dichlorophenyl)ethyl]-4-methylpiperazine dihydrochloride
Molecular Weight (g/mol):
346.12
Molecular Formula:
C13H20Cl4N2
InChi Key:
NXFDBTLQOARIMH-UHFFFAOYSA-N
SMILES:
Cl.Cl.CN1CCN(CCC2=CC(Cl)=C(Cl)C=C2)CC1
1
–
1
of
1
results
1
R&D Systems™ BD 1063 dihydrochloride
CAS: 206996-13-6 Molecular Formula: C13H20Cl4N2