accessibility menu, dialog, popup

UserName

Viewing profile as user:
HNNOHNNOOOHOOH(E/Z)

CAS RN 207572-68-7

IUPAC Name: bis(N-(dicyclopropylmethyl)-4,5-dihydro-1,3-oxazol-2-amine); but-2-enedioic acid
Synonyms: butenedioic acid; bis(hyperium)
Molecular Weight (g/mol): 476.57
Molecular Formula: C24H36N4O6
InChi Key: LZFATBMLSYHRTC-UHFFFAOYSA-N
SMILES: OC(=O)C=CC(O)=O.C1CC1C(NC1=NCCO1)C1CC1.C1CC1C(NC1=NCCO1)C1CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results