CAS RN 207801-28-3
CAS RN 207801-28-3
IUPAC Name:
yttrium(3+) triacetate
Synonyms:
yttrium(3+) triacetate
Molecular Weight (g/mol):
266.04
Molecular Formula:
C6H9O6Y
InChi Key:
JUWHRRAPBUAYTA-UHFFFAOYSA-K
SMILES:
[Y+3].CC([O-])=O.CC([O-])=O.CC([O-])=O
1
–
1
of
1
results
1
Thermo Scientific Chemicals Yttrium acetate hydrate, 99.9%, (trace metal basis)
CAS: 207801-28-3 Molecular Formula: C6H9O6Y Molecular Weight (g/mol): 266.04 MDL Number: MFCD00150128 InChI Key: JUWHRRAPBUAYTA-UHFFFAOYSA-K Synonym: yttriumacetatetetrahydrate,yttrium acetate-water 1/3/4,yttrium 3+ triacetate tetrahydrate,yttrium 3+ ion triacetate tetrahydrate PubChem CID: 19818017 SMILES: [Y+3].CC([O-])=O.CC([O-])=O.CC([O-])=O