CAS RN 209783-80-2
IUPAC Name:
(pyridin-3-yl)methyl N-({4-[(2-aminophenyl)carbamoyl]phenyl}methyl)carbamate
Synonyms:
entinostat
Molecular Weight (g/mol):
376.42
Molecular Formula:
C21H20N4O3
InChi Key:
INVTYAOGFAGBOE-UHFFFAOYSA-N
SMILES:
NC1=CC=CC=C1NC(=O)C1=CC=C(CNC(=O)OCC2=CC=CN=C2)C=C1
1
–
2
of
2
results
1
Thermo Scientific Chemicals Entinostat
CAS: 209783-80-2 Molecular Formula: C21H20N4O3
2
Tocris Bioscience™ MS 275
HDAC (Class I) inhibitor