Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ MS 275
GREENER_CHOICESDP

Catalog No. 620850 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
62-085-0 50 mg
62-081-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 62-085-0 Supplier R&D Systems™ Supplier No. 6208/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. HDAC (Class I) inhibitor

MS 275 is a class I HDAC inhibitor (IC50 values are 0.18, 0.74, 44.9 and >100 μM for HDAC1, 3, 8 and 6, respectively). Exhibits antiproliferative effects and induces apoptosis in a range of tumor cell lines in vitro and in vivo. Increases estrogen receptor α- and aromatase expression in breast cancer cells. Inhibits PCB-induced neuronal cell death by preventing HDAC3 binding and histone deacetylation within the synapsin-1 promoter.

Specifications

Chemical Name or Material (Pyridin-3-yl)methyl 4-(2-aminophenylcarbamoyl)benzylcarbamate
CAS 209783-80-2
Assay Percent Range >99%
Quantity 50 mg
Product Line Tocris
Molecular Formula C21H20N4O3
Formula Weight Observed MW: 376.41
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.