accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 212332-35-9

IUPAC Name: [4-(5-ethoxypyridin-3-yl)but-3-en-1-yl](methyl)amine; bis(but-2-enedioic acid)
Synonyms: [4-(5-ethoxypyridin-3-yl)but-3-en-1-yl](methyl)amine; bis(butenedioic acid)
Molecular Weight (g/mol): 438.43
Molecular Formula: C20H26N2O9
InChi Key: GEWVPSJQGJBDLM-UHFFFAOYSA-N
SMILES: OC(=O)C=CC(O)=O.OC(=O)C=CC(O)=O.CCOC1=CN=CC(C=CCCNC)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results