accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 212612-16-3

CH3OHOOPOOCH3OH3COH

CAS RN 212612-16-3

IUPAC Name: 27-ethoxy-26,28-dihydroxypentacyclo[19.3.1.1³,⁷.1⁹,¹³.1¹⁵,¹⁹]octacosa-1(25),3,5,7(28),9,11,13(27),15,17,19(26),21,23-dodecaen-25-yl diethyl phosphate
Synonyms: 27-ethoxy-26,28-dihydroxypentacyclo[19.3.1.1³,⁷.1⁹,¹³.1¹⁵,¹⁹]octacosa-1(25),3,5,7(28),9,11,13(27),15,17,19(26),21,23-dodecaen-25-yl diethyl phosphate
Molecular Weight (g/mol): 588.64
Molecular Formula: C34H37O7P
InChi Key: AYAKMQZSWFERNT-UHFFFAOYSA-N
SMILES: CCOC1=C2CC3=CC=CC(CC4=CC=CC(CC5=CC=CC(CC1=CC=C2)=C5O)=C4OP(=O)(OCC)OCC)=C3O
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results