CAS RN 2127150-20-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ Janelia Fluor 669, SE
Far red dye (Reactive group: NHS ester). Applications: super resolution microscopy e.g. dSTORM & STED, confocal and live cell imaging