accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3HONHNNNHOOHClNNCH3H3C

CAS RN 212844-54-7

IUPAC Name: 2-chloro-4-({2-[(1-hydroxy-3-methylbutan-2-yl)amino]-9-(propan-2-yl)-9H-purin-6-yl}amino)benzoic acid
Synonyms: purvalanol
Molecular Weight (g/mol): 432.91
Molecular Formula: C20H25ClN6O3
InChi Key: ZKDXRFMOHZVXSG-UHFFFAOYNA-N
SMILES: CC(C)C(CO)NC1=NC(NC2=CC(Cl)=C(C=C2)C(O)=O)=C2N=CN(C(C)C)C2=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)