accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3(R)H3CCH3HO(R)(S)(S)(S)OH(R)H3C(S)(R)H3C

CAS RN 2140-46-7

IUPAC Name: (1R,3aS,3bS,7S,9aR,9bS,11aR)-1-[(2R)-6-hydroxy-6-methylheptan-2-yl]-9a,11a-dimethyl-1H,2H,3H,3aH,3bH,4H,6H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-ol
Synonyms: 25-hydroxycholesterol
Molecular Weight (g/mol): 402.66
Molecular Formula: C27H46O2
InChi Key: INBGSXNNRGWLJU-ZHHJOTBYSA-N
SMILES: C[C@H](CCCC(C)(C)O)[C@H]1CC[C@H]2[C@@H]3CC=C4C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]12C
Product Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)