Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ 25-Hydroxycholesterol
GREENER_CHOICESDP

Catalog No. 574110 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
57-411-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 57-411-0 Supplier Tocris Bioscience™ Supplier No. 5741/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Antiviral; also α-crystallin (cryAB) chaperone

25-Hydroxycholesterol is an antiviral. Stimulates expression of miR-185 microRNA which inhibits the hepatitis C virus life cycle via inhibition of cholesterol biosynthesis and virus cell membrane fusion. Also α-crystallin (cryAB) chaperone (Kd = 10.1 μM). Blocks α-crystallin aggregation and partially reverses cataract formation by binding and stabilizing soluble forms of cryAB in vitro and in vivo.

Specifications

CAS 2140-46-7
Quantity 10 mg
Molecular Formula C27H46O2
Formula Weight 402.65
Product Type Inhibitor

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.