accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 214262-98-3

OOF

CAS RN 214262-98-3

IUPAC Name: 1-(3-fluorophenyl)cyclohexane-1-carboxylate
Synonyms: 1-(3-fluorophenyl)cyclohexane-1-carboxylate
Molecular Weight (g/mol): 221.25
Molecular Formula: C13H14FO2
InChi Key: RIQOIHJORXEGFT-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1(CCCCC1)C1=CC=CC(F)=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results