accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH2NNHNH3

CAS RN 214358-33-5

IUPAC Name: 1-amino-1-({[3-(azaniumylmethyl)phenyl]methyl}amino)ethylium
Synonyms: 1-amino-1-({[3-(ammoniomethyl)phenyl]methyl}amino)ethylium
Molecular Weight (g/mol): 179.27
Molecular Formula: C10H17N3
InChi Key: RODUKNYOEVZQPR-UHFFFAOYSA-P
SMILES: C[C+](N)NCC1=CC=CC(C[NH3+])=C1
Form 
  • (1)
Product Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results