accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 214360-59-5

IUPAC Name: 2,6-dimethoxy-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Synonyms: 2,6-dimethoxy-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Molecular Weight (g/mol): 265.12
Molecular Formula: C13H20BNO4
InChi Key: VZGDCNFVMWRPOW-UHFFFAOYSA-N
SMILES: COC1=CC=C(B2OC(C)(C)C(C)(C)O2)C(OC)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results