accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 21443-96-9

NH2NHN

CAS RN 21443-96-9

IUPAC Name: 1H-indazol-7-amine
Synonyms: 1H-indazol-7-amine
Molecular Weight (g/mol): 133.15
Molecular Formula: C7H7N3
InChi Key: OTFFCAGPSWJBDK-UHFFFAOYSA-N
SMILES: NC1=C2NN=CC2=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results