accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3(R)NH2NS

CAS RN 21463-31-0

IUPAC Name: (6R)-6-methyl-5,6-dihydro-4H-1,3-thiazin-2-amine
Synonyms: (6R)-6-methyl-5,6-dihydro-4H-1,3-thiazin-2-amine
Molecular Weight (g/mol): 130.21
Molecular Formula: C5H10N2S
InChi Key: BATVOUKHGLKDGQ-SCSAIBSYSA-N
SMILES: C[C@@H]1CCN=C(N)S1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results