accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOOCH3OCH3

CAS RN 2150-41-6

IUPAC Name: methyl 2,4-dimethoxybenzoate
Synonyms: methyl 2,4-dimethoxybenzoate
Molecular Weight (g/mol): 196.2
Molecular Formula: C10H12O4
InChi Key: IHIJFZWLGPEYPJ-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C(OC)C=C1OC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results