accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHNFNN

CAS RN 215303-72-3

IUPAC Name: 4-[4-(4-fluorophenyl)-1-(3-phenylpropyl)-5-(pyridin-4-yl)-1H-imidazol-2-yl]but-3-yn-1-ol
Synonyms: 4-[4-(4-fluorophenyl)-1-(3-phenylpropyl)-5-(pyridin-4-yl)imidazol-2-yl]but-3-yn-1-ol
Molecular Weight (g/mol): 425.51
Molecular Formula: C27H24FN3O
InChi Key: QSUSKMBNZQHHPA-UHFFFAOYSA-N
SMILES: OCCC#CC1=NC(=C(N1CCCC1=CC=CC=C1)C1=CC=NC=C1)C1=CC=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)