Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ RWJ 67657
GREENER_CHOICESDP

Catalog No. 299910 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
29-991-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 29-991-0 Supplier Tocris Bioscience™ Supplier No. 2999/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent, selective p38α and p38β inhibitor

Selective p38α and p38β inhibitor (IC50 values are 1 and 11 μM respectively) that displays no activity at p38γ, p38δ and a range of other kinases. Potently inhibits TNF-α and IL-1β release (IC50 values are 3 and 11 nM respectively) and suppresses HIV-1 replication in T cells in vitro. Displays cardioprotective and anti-inflammatory activity in vivo. Orally active.

Chemical Identifiers

CAS 215303-72-3
Molecular Formula C27H24FN3O

Specifications

Quantity 10 mg
Formula Weight 425.5
Purity >98%
Molecular Weight (g/mol) 425.507
Chemical Name or Material RWJ 67657
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.