accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COHNOOHOCH3H3CCH3

CAS RN 215307-86-1

IUPAC Name: 4-(1,1,3,3-tetramethyl-2-oxo-2,3-dihydro-1H-indene-5-amido)benzoic acid
Synonyms: 4-(1,1,3,3-tetramethyl-2-oxoindene-5-amido)benzoic acid
Molecular Weight (g/mol): 351.4
Molecular Formula: C21H21NO4
InChi Key: KFBPBWUZXBYJDG-UHFFFAOYSA-N
SMILES: CC1(C)C(=O)C(C)(C)C2=C1C=CC(=C2)C(=O)NC1=CC=C(C=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)