accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NHOHOHNO(E/Z)

CAS RN 215543-92-3

IUPAC Name: 3-{4-methyl-2-[(2-oxo-2,3-dihydro-1H-indol-3-ylidene)methyl]-1H-pyrrol-3-yl}propanoic acid
Synonyms: 3-{4-methyl-2-[(2-oxo-1H-indol-3-ylidene)methyl]-1H-pyrrol-3-yl}propanoic acid
Molecular Weight (g/mol): 296.33
Molecular Formula: C17H16N2O3
InChi Key: JNDVEAXZWJIOKB-UHFFFAOYSA-N
SMILES: CC1=CNC(C=C2C(=O)NC3=CC=CC=C23)=C1CCC(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)