accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2157-24-6

IUPAC Name: 3-(3-aminopropoxy)propan-1-amine
Synonyms: 3-(3-aminopropoxy)propan-1-amine
Molecular Weight (g/mol): 132.21
Molecular Formula: C6H16N2O
InChi Key: KRPRVQWGKLEFKN-UHFFFAOYSA-N
SMILES: NCCCOCCCN

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results