accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2NNHSSOHOO

CAS RN 21668-81-5

IUPAC Name: 3-(carbamimidoylsulfanyl)propane-1-sulfonic acid
Synonyms: 3-(carbamimidoylsulfanyl)propane-1-sulfonic acid
Molecular Weight (g/mol): 198.26
Molecular Formula: C4H10N2O3S2
InChi Key: MQLJIOAPXLAGAP-UHFFFAOYSA-N
SMILES: NC(=N)SCCCS(O)(=O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results