Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(Carbamimidoylthio)-1-propanesulfonic Acid 98.0+%, TCI America™
SDP

Catalog No. C298225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C298225G 25 g
C2982100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C298225G Supplier TCI America Supplier No. C298225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 21668-81-5
Molecular Formula C4H10N2O3S2
Molecular Weight (g/mol) 198.255
MDL Number MFCD00085942
InChI Key MQLJIOAPXLAGAP-UHFFFAOYSA-N
Synonym 3-amidinothio-1-propanesulfonic acid, 3-s-isothiuronium propyl sulfonate, 3-carbamimidoylthio propane-1-sulfonic acid, 3-carbamimidoylsulfanyl propane-1-sulfonic acid, 3-amino-iminomethyl-thio-1-propanesulfonic acid, 3-carbamimidoylthio-1-propanesulfonic acid, 3-aminoiminomethyl thio-1-propanesulfonic acid, 3-aminoiminomethyl thio propanesulphonic acid, 1-propanesulfonic acid,3-aminoiminomethyl thio
PubChem CID 89008
IUPAC Name 3-carbamimidoylsulfanylpropane-1-sulfonic acid
SMILES C(CSC(=N)N)CS(=O)(=O)O

Specifications

Melting Point 257°C
Color White
Quantity 25 g
UN Number 3261
Formula Weight 198.26
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 3-(Carbamimidoylthio)-1-propanesulfonic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.