accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COONH2HNONHNH2

CAS RN 217480-27-8

IUPAC Name: 2-aminopropyl 6-amino-2-{2-[2-(naphthalen-2-yl)-1H-benzo[g]indol-3-yl]acetamido}hexanoate
Synonyms: 2-aminopropyl 6-amino-2-{2-[2-(naphthalen-2-yl)-1H-benzo[g]indol-3-yl]acetamido}hexanoate
Molecular Weight (g/mol): 536.68
Molecular Formula: C33H36N4O3
InChi Key: NFVRGDRCCNEGBS-UHFFFAOYNA-N
SMILES: CC(N)COC(=O)C(CCCCN)NC(=O)CC1=C(NC2=C1C=CC1=CC=CC=C21)C1=CC=C2C=CC=CC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results