accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNOOClHNOH2NF

CAS RN 217645-70-0

IUPAC Name: [5-chloro-2-(2-{4-[(4-fluorophenyl)methyl]-2-methylpiperazin-1-yl}-2-oxoethoxy)phenyl]urea
Synonyms: 5-chloro-2-(2-{4-[(4-fluorophenyl)methyl]-2-methylpiperazin-1-yl}-2-oxoethoxy)phenylurea
Molecular Weight (g/mol): 434.9
Molecular Formula: C21H24ClFN4O3
InChi Key: XQYASZNUFDVMFH-UHFFFAOYNA-N
SMILES: CC1CN(CC2=CC=C(F)C=C2)CCN1C(=O)COC1=C(NC(N)=O)C=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)