Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ BX 471
GSA_VAGREENER_CHOICESDP

Catalog No. 349650 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
34-965-0 50 mg
34-961-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 34-965-0 Supplier R&D Systems™ Supplier No. 3496/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent, selective CCR1 antagonist

Potent, selective non-peptide CCR1 antagonist (Ki = 1 nM for human CCR1). Exhibits 250-fold selectivity for CCR1 over CCR2, CCR5 and CXCR4. Inhibits MIP-α/CCL3-induced intracellular Ca2+ mobilization. Orally active; effectively reduces disease severity in a rat model of multiple sclerosis. Decreases renal fibrosis in a mouse model of obstructive nephropathy.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Chemical Identifiers

CAS 217645-70-0
Molecular Formula C21H24ClFN4O3

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 434.89
Purity >98%
Molecular Weight (g/mol) 434.896
Chemical Name or Material BX 471
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.