accessibility menu, dialog, popup

UserName

Viewing profile as user:
HNOHNONHOS

CAS RN 219766-25-3

IUPAC Name: N-{2-[(2-oxoazepan-3-yl)carbamoyl]phenyl}-1-benzothiophene-2-carboxamide
Synonyms: N-{2-[(2-oxoazepan-3-yl)carbamoyl]phenyl}-1-benzothiophene-2-carboxamide
Molecular Weight (g/mol): 407.49
Molecular Formula: C22H21N3O3S
InChi Key: TUSCYCAIGRVBMD-UHFFFAOYNA-N
SMILES: O=C(NC1=CC=CC=C1C(=O)NC1CCCCNC1=O)C1=CC2=CC=CC=C2S1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results