accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 22041-19-6

H3COONH

CAS RN 22041-19-6

IUPAC Name: 1-(2-ethoxy-2-oxoethyl)pyrrolidin-1-ium
Synonyms: 1-(2-ethoxy-2-oxoethyl)pyrrolidin-1-ium
Molecular Weight (g/mol): 158.22
Molecular Formula: C8H16NO2
InChi Key: IIIBKVMTKMLDFJ-UHFFFAOYSA-O
SMILES: CCOC(=O)C[NH+]1CCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results