accessibility menu, dialog, popup

UserName

Viewing profile as user:
IBrOHBrOHN(E/Z)

CAS RN 220904-83-6

IUPAC Name: 3-[(3,5-dibromo-4-hydroxyphenyl)methylidene]-5-iodo-2,3-dihydro-1H-indol-2-one
Synonyms: 3-[(3,5-dibromo-4-hydroxyphenyl)methylidene]-5-iodo-1H-indol-2-one
Molecular Weight (g/mol): 520.95
Molecular Formula: C15H8Br2INO2
InChi Key: LMXYVLFTZRPNRV-UHFFFAOYSA-N
SMILES: OC1=C(Br)C=C(C=C2C(=O)NC3=C2C=C(I)C=C3)C=C1Br
Form 
  • (1)
Product Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results