accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNH3CNNNCH3CH3H3CCH3H3CH3CHCl(?)

CAS RN 220953-69-5

IUPAC Name: N-butyl-N-ethyl-2,5,6-trimethyl-7-(2,4,6-trimethylphenyl)-7H-pyrrolo[2,3-d]pyrimidin-4-amine hydrochloride
Synonyms: antalarmin hydrochloride
Molecular Weight (g/mol): 415.02
Molecular Formula: C24H35ClN4
InChi Key: CGDGXEDXEXACKQ-UHFFFAOYSA-N
SMILES: Cl.CCCCN(CC)C1=C2C(C)=C(C)N(C2=NC(C)=N1)C1=C(C)C=C(C)C=C1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)