accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3H3CNNNH2NN

CAS RN 221243-82-9

IUPAC Name: 1-tert-butyl-3-(naphthalen-1-yl)-1H-pyrazolo[3,4-d]pyrimidin-4-amine
Synonyms: 1-tert-butyl-3-(naphthalen-1-yl)pyrazolo[3,4-d]pyrimidin-4-amine
Molecular Weight (g/mol): 317.4
Molecular Formula: C19H19N5
InChi Key: XSHQBIXMLULFEV-UHFFFAOYSA-N
SMILES: CC(C)(C)N1N=C(C2=C(N)N=CN=C12)C1=C2C=CC=CC2=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)