CAS RN 2222635-15-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ DCLK1-IN-1
Potent and selective doublecortin like kinase 1/2 (DCLK1/2) inhibitor; orally bioavailable