CAS RN 22260-51-1
IUPAC Name:
(4R,7R)-10-bromo-N-[(1S,2S,4R,7S)-2-hydroxy-7-(2-methylpropyl)-5,8-dioxo-4-(propan-2-yl)-3-oxa-6,9-diazatricyclo[7.3.0.0²,⁶]dodecan-4-yl]-6-methyl-6,11-diazatetracyclo[7.6.1.0²,⁷.0¹²,¹⁶]hexadeca-1(16),2,9,12,14-pentaene-4-carboxamide; methanesulfonic acid
Synonyms:
parlodel bromocriptine mesilate pravidel bromocriptine methanesulfonate
Molecular Weight (g/mol):
750.71
Molecular Formula:
C33H44BrN5O8S
InChi Key:
NOJMTMIRQRDZMT-GSPXQYRGSA-N
SMILES:
CS(O)(=O)=O.CC(C)C[C@@H]1N2C(=O)[C@](NC(=O)[C@H]3CN(C)[C@@H]4CC5=C(Br)NC6=CC=CC(=C56)C4=C3)(O[C@@]2(O)[C@@H]2CCCN2C1=O)C(C)C
1
–
1
of
1
results
1
R&D Systems™ Bromocriptine mesylate
CAS: 22260-51-1 Molecular Formula: C33H44BrN5O8S