CAS RN 2227372-56-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ Loreclezole hydrochloride
CAS: 2227372-56-5 Molecular Formula: C10H7Cl4N3