accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SNNOHNNNFFFFFF

CAS RN 223499-30-7

IUPAC Name: N-{4-[3,5-bis(trifluoromethyl)-1H-pyrazol-1-yl]phenyl}-4-methyl-1,2,3-thiadiazole-5-carboxamide
Synonyms: N-{4-[3,5-bis(trifluoromethyl)pyrazol-1-yl]phenyl}-4-methyl-1,2,3-thiadiazole-5-carboxamide
Molecular Weight (g/mol): 421.32
Molecular Formula: C15H9F6N5OS
InChi Key: XPRZIORDEVHURQ-UHFFFAOYSA-N
SMILES: CC1=C(SN=N1)C(=O)NC1=CC=C(C=C1)N1N=C(C=C1C(F)(F)F)C(F)(F)F
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results