CAS RN 2244989-34-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ B 973B
Positive allosteric modulator of α7 nAChRs; analgesic