Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ B 973B
GREENER_CHOICE

Catalog No. 676450
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
67-645-0 50 mg
67-641-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 67-645-0 Supplier Tocris Bioscience™ Supplier No. 6764/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Positive allosteric modulator of α7 nAChRs; analgesic

B 973B is a positive allosteric modulator of α7 nAChRs (EC50 ∽0.3 μM); active enantiomer. Increases potency of acetylcholine (EC50 values are 0.007 mM and 0.3 mM in the presence and absence of B 937B, respectively). Potentiates peak acetylcholine-induced currents 6-fold relative to maximal acetylcholine, and slows channel desensitization, resulting in a 6900-fold increase in charge transfer. Decreases paw edema in vivo. Analgesic.

Specifications

Chemical Name or Material 3,4-Difluoro-N-[(1S)-1-[6-[4-(2-pyridinyl)-1-piperazinyl]-2-pyrazinyl]ethyl]benzenepropanamide
CAS 2244989-34-0
Quantity 50 mg
Molecular Formula C24H26F2N6O
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.