CAS RN 2245697-86-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Thalidomide 4'-oxyacetamide-alkylC4-amine
PROTACs (Proteolysis Targeting Chimeras)