accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 22483-09-6

IUPAC Name: 2,2-dimethoxyethan-1-amine
Synonyms: 2,2-dimethoxyethanamine
Molecular Weight (g/mol): 105.14
Molecular Formula: C4H11NO2
InChi Key: QKWWDTYDYOFRJL-UHFFFAOYSA-N
SMILES: COC(CN)OC
Molecular Weight (g/mol) 
  • (6)
  • (6)
Percent Purity 
  • (2)
  • (1)
  • (9)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (3)
  • (3)
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (3)
  • (2)
  • (7)
  • (6)
  • (6)
  • (2)
  • (1)
  • (9)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)
  • (3)
  • (3)
  • (2)
  • (6)